siRNA Details
Gene Symbol: LINC00544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033889.1 | 4 | 438 | |||
GenBank mRNA | BC132810.1 | - | 2 | 169 | ||
BC132836.1 | - | 2 | 169 | |||
BX118434.1 | - | 4 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
