siRNA Details
Gene Symbol: KIAA0087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022006.1 | 2 | 1369 | |||
GenBank mRNA | BC128257.1 | - | 2 | 1162 | ||
D42038.1 | - | 2 | 1331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
