siRNA Details
Gene Symbol: PHF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304333.2 | NP_001291262.1 | 8 | 1597 | ||
NM_015297.2 | NP_056112.1 | 8 | 1593 | |||
NR_136307.1 | 8 | 1597 | ||||
NR_136308.1 | 8 | 1593 | ||||
XM_011517827.1 | XP_011516129.1 | 9 | 1585 | |||
XM_011517828.2 | XP_011516130.1 | 8 | 1689 | |||
XM_011517829.1 | XP_011516131.1 | 8 | 1847 | |||
XM_017014553.1 | XP_016870042.1 | 10 | 1879 | |||
XM_017014554.1 | XP_016870043.1 | 10 | 1659 | |||
XM_017014555.1 | XP_016870044.1 | 10 | 1631 | |||
XM_017014556.1 | XP_016870045.1 | 10 | 2163 | |||
GenBank mRNA | AB028968.1 | - | 8 | 1462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
