siRNA Details
Gene Symbol: KIAA1549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164665.1 | NP_001158137.1 | 4 | 3064 | ||
NM_020910.2 | NP_065961.2 | 4 | 3064 | |||
XM_011516442.2 | XP_011514744.1 | 5 | 3291 | |||
GenBank mRNA | AB046769.2 | - | 3 | 2808 | ||
AM989468.1 | - | 4 | 3064 | |||
AM989469.1 | - | 4 | 3064 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
