siRNA Details
Gene Symbol: FAM149B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173348.1 | NP_775483.1 | 6 | 910 | ||
XM_005269744.2 | XP_005269801.1 | 6 | 910 | |||
XM_005269745.2 | XP_005269802.1 | 6 | 910 | |||
XM_005269747.3 | XP_005269804.1 | 6 | 701 | |||
XM_011539737.2 | XP_011538039.1 | 6 | 713 | |||
XM_011539740.2 | XP_011538042.1 | 6 | 910 | |||
XM_017016164.1 | XP_016871653.1 | 6 | 714 | |||
XM_017016165.1 | XP_016871654.1 | 5 | 557 | |||
XM_017016166.1 | XP_016871655.1 | 5 | 594 | |||
XM_017016167.1 | XP_016871656.1 | 6 | 910 | |||
GenBank mRNA | AB023191.1 | - | 5 | 638 | ||
AK023439.1 | - | 3 | 95 | |||
AK303436.1 | - | 6 | 674 | |||
BC015394.1 | - | 4 | 511 | 4431919 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
