siRNA Details
Gene Symbol: GCNT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080615.1 | NP_542182.1 | 5 | 1492 | ||
GenBank mRNA | AK131203.1 | - | 5 | 1492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
