siRNA Details
Gene Symbol: ZNF365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014951.2 | NP_055766.2 | 5 | 2863 | ||
GenBank mRNA | AB020651.1 | - | 5 | 2824 | ||
BC035049.1 | - | 4 | 1977 | |||
BC060817.1 | - | 4 | 1972 | |||
BC070073.1 | - | 5 | 2647 | |||
BX647853.1 | - | 5 | 2655 | |||
BX647904.1 | - | 5 | 2655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
