siRNA Details
Gene Symbol: ZNF831 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178457.2 | NP_848552.1 | 5 | 4457 | ||
XM_005260272.3 | XP_005260329.1 | 6 | 5396 | |||
XM_005260273.3 | XP_005260330.1 | 7 | 7470 | |||
XM_006723698.3 | XP_006723761.1 | 6 | 4793 | |||
XM_011528534.2 | XP_011526836.1 | 7 | 6037 | |||
XM_011528536.2 | XP_011526838.1 | 6 | 5219 | |||
XM_011528537.2 | XP_011526839.1 | 7 | 5480 | |||
XM_011528538.2 | XP_011526840.1 | 6 | 5562 | |||
XM_017027642.1 | XP_016883131.1 | 6 | 5851 | |||
XM_017027643.1 | XP_016883132.1 | 7 | 4877 | |||
XM_017027644.1 | XP_016883133.1 | 7 | 4915 | |||
GenBank mRNA | AL713683.1 | - | 1 | 41 | ||
AL833682.1 | - | 5 | 3324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
