siRNA Details
Gene Symbol: PAIP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020459.1 | NP_065192.1 | 4 | 3395 | ||
XM_005264310.3 | XP_005264367.1 | 5 | 3325 | |||
XM_005264311.4 | XP_005264368.1 | 4 | 3371 | |||
XM_011532842.2 | XP_011531144.1 | 4 | 3365 | |||
XM_011532843.2 | XP_011531145.1 | 4 | 3367 | |||
GenBank mRNA | AB032981.1 | - | 4 | 3375 | ||
AL110211.1 | - | 1 | 2403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
