siRNA Details
Gene Symbol: PINLYP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193621.1 | NP_001180550.1 | 5, 6 | 676 | ||
NM_001193622.1 | NP_001180551.1 | 3, 4 | 336 | |||
NM_001321124.1 | NP_001308053.1 | 3, 4 | 356 | |||
XM_011526970.2 | XP_011525272.1 | 4, 5 | 2272 | |||
XM_011526971.2 | XP_011525273.1 | 3, 4 | 698 | |||
GenBank mRNA | BC071811.1 | - | 3, 4 | 336 | ||
CN355575.1 | - | 3, 4 | 315 | |||
HY005068.1 | - | 3, 4 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
