siRNA Details
Gene Symbol: ZNF391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076781.2 | NP_001070249.1 | 3 | 755 | ||
NM_001322288.1 | NP_001309217.1 | 4 | 979 | |||
NM_001322289.1 | NP_001309218.1 | 4 | 834 | |||
NM_001322293.1 | NP_001309222.1 | 3 | 723 | |||
XM_005249082.3 | XP_005249139.1 | 3 | 540 | |||
XM_006715086.2 | XP_006715149.1 | 3 | 481 | |||
XM_011514569.1 | XP_011512871.1 | 3 | 463 | |||
GenBank mRNA | AK294965.1 | - | 3 | 714 | ||
U80232.1 | - | 1 | 47 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
