siRNA Details
Gene Symbol: PPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026569.1 | - | 1 | 31 | ||
AK290085.1 | - | 1 | 32 | |||
AK290851.1 | - | 1 | 34 | |||
AK297340.1 | - | 1 | 31 | |||
AK316564.1 | - | 1 | 31 | |||
AU100120.1 | - | 1 | 70 | |||
BC093076.1 | - | 1 | 31 | 30414589 | ||
BC106030.1 | - | 1 | 23 | 2986147 | ||
BC137058.1 | - | 1 | 76 | 9021055 | ||
EU794650.1 | - | 1 | 23 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
