siRNA Details
Gene Symbol: RPH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143854.1 | NP_001137326.1 | 22 | 4029 | ||
NM_014954.3 | NP_055769.2 | 21 | 4017 | |||
XM_011538048.1 | XP_011536350.1 | 22 | 3833 | |||
XM_017019034.1 | XP_016874523.1 | 14 | 3287 | |||
GenBank mRNA | AB023202.1 | - | 22 | 3950 | ||
BX647281.1 | - | 14 | 4622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
