siRNA Details
Gene Symbol: RAD21L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136566.2 | NP_001130038.2 | 2 | 180 | ||
XM_006723602.2 | XP_006723665.1 | 2 | 217 | |||
XM_006723603.2 | XP_006723666.1 | 2 | 383 | |||
XM_006723604.2 | XP_006723667.1 | 2 | 217 | |||
XM_006723605.3 | XP_006723668.1 | 2 | 217 | |||
XM_011529317.2 | XP_011527619.1 | 2 | 379 | |||
GenBank mRNA | BC157891.1 | - | 2 | 180 | 8862712 | |
BC171911.1 | - | 2 | 180 | 9054565 | ||
HQ603827.1 | - | 1 | 95 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
