siRNA Details
Gene Symbol: ARSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524545.2 | XP_011522847.1 | 11 | 3940 | ||
XM_017024368.1 | XP_016879857.1 | 11 | 3744 | |||
GenBank mRNA | AB023218.1 | - | 11 | 3672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
