siRNA Details
Gene Symbol: RUFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291994.1 | NP_001278923.1 | 14 | 3862 | ||
NM_014961.4 | NP_055776.1 | 13 | 3808 | |||
XM_011531755.2 | XP_011530057.1 | 13 | 3422 | |||
XM_017007891.1 | XP_016863380.1 | 14 | 3154 | |||
XM_017007892.1 | XP_016863381.1 | 13 | 3122 | |||
GenBank mRNA | AB020678.1 | - | 13 | 3614 | ||
AK021769.1 | - | 2 | 1629 | |||
BC051716.1 | - | 13 | 3676 | 5265230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
