siRNA Details
Gene Symbol: AHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029882.3 | NP_001025053.1 | 6 | 5629 | ||
XM_005245848.3 | XP_005245905.1 | 7 | 5864 | |||
XM_005245849.3 | XP_005245906.1 | 7 | 5969 | |||
XM_005245850.3 | XP_005245907.1 | 8 | 5989 | |||
XM_005245851.3 | XP_005245908.1 | 7 | 5861 | |||
XM_005245852.3 | XP_005245909.1 | 6 | 5638 | |||
XM_011541255.1 | XP_011539557.1 | 7 | 5543 | |||
XM_011541256.2 | XP_011539558.1 | 7 | 5983 | |||
XM_011541257.2 | XP_011539559.1 | 6 | 6738 | |||
GenBank mRNA | AK125431.1 | - | 1 | 2347 | ||
BC002677.2 | - | 1 | 1425 | |||
BC014394.1 | - | 1 | 1425 | |||
U79259.1 | - | 1 | 1390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
