siRNA Details
Gene Symbol: ZNF484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007101.2 | NP_001007102.1 | 4 | 631 | ||
NM_001261458.1 | NP_001248387.1 | 4 | 701 | |||
NM_001261459.1 | NP_001248388.1 | 6 | 772 | |||
NM_001261460.1 | NP_001248389.1 | 6 | 702 | |||
NM_031486.2 | NP_113674.1 | 5 | 676 | |||
XM_006717299.3 | XP_006717362.1 | 5 | 697 | |||
XM_011519073.2 | XP_011517375.1 | 6 | 813 | |||
XM_011519074.2 | XP_011517376.1 | 5 | 586 | |||
XM_011519075.2 | XP_011517377.1 | 6 | 742 | |||
XM_017015178.1 | XP_016870667.1 | 7 | 768 | |||
XM_017015179.1 | XP_016870668.1 | 7 | 616 | |||
XM_017015180.1 | XP_016870669.1 | 5 | 546 | |||
XM_017015181.1 | XP_016870670.1 | 7 | 780 | |||
GenBank mRNA | AK091203.1 | - | 4 | 581 | ||
AK292997.1 | - | 7 | 760 | |||
AK299274.1 | - | 4 | 599 | |||
AK309617.1 | - | 5 | 676 | |||
AL833202.1 | - | 5 | 646 | |||
BC112394.1 | - | 4 | 526 | 40008083 | ||
BC112396.1 | - | 4 | 526 | 40008084 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
