siRNA Details
Gene Symbol: ANKRD19P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026868.1 | 10 | 2162 | |||
GenBank mRNA | AK093497.1 | - | 8 | 1754 | ||
BC038951.1 | - | 9 | 2085 | 5733799 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
