siRNA Details
Gene Symbol: GNAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102386.2 | NP_001095856.1 | 3 | 328 | ||
XM_011516124.2 | XP_011514426.1 | 3 | 822 | |||
GenBank mRNA | BC147016.1 | - | 3 | 328 | 9056944 | |
BC147017.1 | - | 3 | 328 | 9056945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
