siRNA Details
Gene Symbol: LRFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020862.1 | NP_065913.1 | 2 | 1678 | ||
XM_005259102.4 | XP_005259159.1 | 5 | 1878 | |||
XM_017027033.1 | XP_016882522.1 | 5 | 1993 | |||
GenBank mRNA | AB040917.1 | - | 2 | 1467 | ||
BC014678.1 | - | 774 | ||||
BC025310.1 | - | 2 | 991 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
