siRNA Details
Gene Symbol: C22orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302819.1 | NP_001289748.1 | 1 | 439 | ||
NM_001302820.1 | NP_001289749.1 | 1 | 439 | |||
NM_015372.2 | NP_056187.1 | 2 | 230 | |||
NR_126507.1 | 2 | 230 | ||||
GenBank mRNA | AJ704765.1 | - | 2 | 181 | ||
AJ704766.1 | - | 2 | 181 | |||
AK294232.1 | - | 1 | 409 | |||
AK315264.1 | - | 2 | 228 | |||
AL050256.1 | - | 2 | 218 | |||
DA485290.1 | - | 1 | 440 | |||
DB102820.1 | - | 2 | 230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
