siRNA Details
Gene Symbol: DNAJC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129887.1 | NP_001123359.1 | 11 | 1398 | ||
NM_014377.1 | NP_055192.1 | 13 | 1557 | |||
XM_005250269.2 | XP_005250326.1 | 14 | 1506 | |||
XM_011516028.2 | XP_011514330.1 | 13 | 1957 | |||
XM_011516029.2 | XP_011514331.1 | 14 | 1418 | |||
XM_011516030.2 | XP_011514332.1 | 9 | 758 | |||
XM_017011968.1 | XP_016867457.1 | 14 | 1593 | |||
GenBank mRNA | AK126382.1 | - | 1 | 402 | ||
AL832847.1 | - | 12 | 1341 | |||
BC000859.1 | - | 5 | 401 | |||
BC125057.1 | - | 12 | 1249 | |||
BC139751.1 | - | 13 | 1305 | |||
X98260.1 | - | 11 | 1191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
