siRNA Details
Gene Symbol: DNAJC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129887.1 | NP_001123359.1 | 14, 15 | 1894 | ||
NM_014377.1 | NP_055192.1 | 16, 17 | 2053 | |||
XM_005250269.2 | XP_005250326.1 | 17, 18 | 2002 | |||
XM_011516028.2 | XP_011514330.1 | 16, 17 | 2453 | |||
XM_011516029.2 | XP_011514331.1 | 17, 18 | 1914 | |||
XM_011516030.2 | XP_011514332.1 | 12, 13 | 1254 | |||
XM_017011968.1 | XP_016867457.1 | 17, 18 | 2089 | |||
GenBank mRNA | AK126382.1 | - | 2 | 2058 | ||
AL832847.1 | - | 15, 16 | 1837 | |||
BC000859.1 | - | 8, 9 | 897 | |||
BC125057.1 | - | 15, 16 | 1745 | |||
BC139751.1 | - | 16, 17 | 1801 | |||
X98260.1 | - | 14, 15 | 1687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
