siRNA Details
Gene Symbol: STRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020704.2 | NP_065755.1 | 21 | 2546 | ||
XM_006716069.3 | XP_006716132.1 | 20 | 2341 | |||
XM_011516432.2 | XP_011514734.1 | 18 | 2194 | |||
XM_017012471.1 | XP_016867960.1 | 15 | 1869 | |||
XM_017012472.1 | XP_016867961.1 | 19 | 4011 | |||
GenBank mRNA | AB032996.1 | - | 21 | 2520 | ||
AK290165.1 | - | 21 | 2546 | |||
AK302339.1 | - | 13 | 1751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
