siRNA Details
Gene Symbol: TRAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042646.2 | NP_001036111.1 | 16 | 4037 | ||
XM_005264956.2 | XP_005265013.1 | 16 | 3782 | |||
XM_005264958.2 | XP_005265015.1 | 15 | 3900 | |||
XM_005264960.3 | XP_005265017.1 | 17 | 3938 | |||
XM_017005906.1 | XP_016861395.1 | 18 | 4596 | |||
XM_017005907.1 | XP_016861396.1 | 15 | 3960 | |||
XM_017005908.1 | XP_016861397.1 | 15 | 4038 | |||
XM_017005909.1 | XP_016861398.1 | 17 | 4481 | |||
XM_017005910.1 | XP_016861399.1 | 15 | 3837 | |||
XM_017005911.1 | XP_016861400.1 | 17 | 4280 | |||
XM_017005913.1 | XP_016861402.1 | 17 | 3875 | |||
GenBank mRNA | AB028965.1 | - | 16 | 4037 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
