siRNA Details
Gene Symbol: FEZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024613.3 | NP_001019784.2 | 1 | 661 | ||
XM_005250337.3 | XP_005250394.1 | 2 | 2332 | |||
GenBank mRNA | AK300742.1 | - | 1 | 790 | ||
AY726588.1 | - | 1 | 582 | |||
BC127714.1 | - | 1 | 649 | 40127896 | ||
BC136684.1 | - | 1 | 717 | 9020674 | ||
BC144367.1 | - | 1 | 717 | 9052888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
