siRNA Details
Gene Symbol: KNSTRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142761.1 | NP_001136233.1 | 2 | 359 | ||
NM_001142762.1 | NP_001136234.1 | 2 | 359 | |||
NM_033286.3 | NP_150628.3 | 2 | 359 | |||
GenBank mRNA | AK027408.1 | - | 2 | 101 | ||
AK125765.1 | - | 2 | 359 | |||
AK298156.1 | - | 2 | 85 | |||
AK301887.1 | - | 2 | 359 | |||
AY652615.1 | - | 2 | 258 | |||
BC004543.2 | - | 2 | 68 | |||
BC014060.2 | - | 2 | 68 | |||
BC045739.1 | - | 2 | 140 | |||
BC064344.1 | - | 2 | 67 | |||
BC107802.1 | - | 2 | 125 | |||
BC118559.1 | - | 2 | 288 | 40023827 | ||
BC118639.1 | - | 2 | 288 | 40023825 | ||
DC396708.1 | - | 2 | 359 | |||
U81002.1 | - | 2 | 115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
