siRNA Details
Gene Symbol: KNSTRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142761.1 | NP_001136233.1 | 9 | 1606 | ||
NM_001142762.1 | NP_001136234.1 | 8 | 1268 | |||
NM_033286.3 | NP_150628.3 | 9 | 1330 | |||
GenBank mRNA | AK125765.1 | - | 9 | 1287 | ||
AK298156.1 | - | 8 | 994 | |||
AK301887.1 | - | 9 | 1330 | |||
BC004543.2 | - | 9 | 1039 | |||
BC014060.2 | - | 9 | 1039 | |||
BC045739.1 | - | 9 | 1111 | |||
BC064344.1 | - | 9 | 1038 | |||
BC107802.1 | - | 9 | 1096 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
