siRNA Details
Gene Symbol: NGDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042635.1 | NP_001036100.1 | 9, 10 | 905 | ||
NM_015514.1 | NP_056329.1 | 9, 10 | 905 | |||
GenBank mRNA | AK022215.1 | - | 6, 7 | 1470 | ||
AK291875.1 | - | 9, 10 | 898 | |||
AL050003.1 | - | 9, 10 | 893 | |||
BC030817.1 | - | 9, 10 | 891 | 4768089 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
