siRNA Details
Gene Symbol: PSMB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099780.1 | NP_001093250.1 | 1 | 1123 | ||
GenBank mRNA | AK122797.1 | - | 1 | 1123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
