siRNA Details
Gene Symbol: GCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006836.1 | NP_006827.1 | 44 | 5784 | ||
GenBank mRNA | BC153881.1 | - | 44 | 5784 | 8860118 | |
D86973.2 | - | 44 | 5784 | |||
U77700.1 | - | 22 | 3249 | |||
U88836.1 | - | 5 | 742 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
