siRNA Details
Gene Symbol: SEC14L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014692.1 | NP_055507.1 | 16 | 4279 | ||
GenBank mRNA | AB007880.2 | - | 16 | 4279 | ||
BC045685.1 | - | 1 | 1998 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
