siRNA Details
Gene Symbol: CEP170B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112726.2 | NP_001106197.1 | 16 | 4423 | ||
NM_015005.2 | NP_055820.2 | 15 | 4396 | |||
XM_005267550.3 | XP_005267607.1 | 16 | 4510 | |||
XM_011536665.2 | XP_011534967.1 | 17 | 4663 | |||
XM_011536666.2 | XP_011534968.1 | 16 | 5026 | |||
XM_011536667.2 | XP_011534969.1 | 9 | 2702 | |||
XM_017021227.1 | XP_016876716.1 | 15 | 5275 | |||
XM_017021228.1 | XP_016876717.1 | 15 | 5170 | |||
GenBank mRNA | AB006622.3 | - | 16 | 4423 | ||
AK091980.1 | - | 4 | 2032 | |||
AK300834.1 | - | 2 | 184 | |||
BC047913.1 | - | 2 | 91 | |||
BC112928.1 | - | 15 | 4427 | 6141521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
