siRNA Details
Gene Symbol: CEP170B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112726.2 | NP_001106197.1 | 19 | 6590 | ||
NM_015005.2 | NP_055820.2 | 18 | 6563 | |||
XM_005267550.3 | XP_005267607.1 | 19 | 6677 | |||
XM_011536665.2 | XP_011534967.1 | 20 | 6830 | |||
XM_011536666.2 | XP_011534968.1 | 19 | 7193 | |||
XM_011536667.2 | XP_011534969.1 | 12 | 4869 | |||
XM_017021227.1 | XP_016876716.1 | 18 | 7442 | |||
XM_017021228.1 | XP_016876717.1 | 18 | 7337 | |||
GenBank mRNA | AA244396.1 | - | 1 | 201 | ||
AB006622.3 | - | 19 | 6591 | |||
AK025023.1 | - | 4 | 2166 | |||
AK091980.1 | - | 8 | 2400 | |||
BC047913.1 | - | 5 | 2259 | |||
BC112928.1 | - | 18 | 6595 | 6141521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
