siRNA Details
Gene Symbol: KHNYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290256.1 | NP_001277185.1 | 8 | 2942 | ||
NM_001290257.1 | NP_001277186.1 | 8 | 2877 | |||
NM_015299.2 | NP_056114.1 | 8 | 3005 | |||
XM_005267474.4 | XP_005267531.1 | 8 | 2961 | |||
XM_011536590.2 | XP_011534892.1 | 8 | 3111 | |||
GenBank mRNA | AB002321.1 | - | 8 | 2942 | ||
BC015780.1 | - | 6 | 2028 | |||
BC042554.1 | - | 8 | 2970 | 5173707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
