siRNA Details
Gene Symbol: AKR1B15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080538.2 | NP_001074007.2 | 10 | 1179 | ||
XM_011516241.2 | XP_011514543.2 | 8 | 1581 | |||
XM_017012224.1 | XP_016867713.1 | 8 | 932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
