siRNA Details
Gene Symbol: TXLNGY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045128.1 | 1 | 107 | |||
NR_045129.1 | 1 | 107 | ||||
GenBank mRNA | AF332224.1 | - | 1 | 83 | ||
AK127387.1 | - | 1 | 115 | |||
BM263181.1 | - | 1 | 107 | |||
DA956406.1 | - | 1 | 118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
