siRNA Details
Gene Symbol: C20orf194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009984.2 | NP_001009984.1 | 10 | 1049 | ||
XM_005260684.3 | XP_005260741.1 | 10 | 1046 | |||
XM_017027783.1 | XP_016883272.1 | 7 | 670 | |||
GenBank mRNA | AK226100.1 | - | 5 | 473 | ||
AK303375.1 | - | 1 | 421 | |||
AK310540.1 | - | 10 | 1067 | |||
AL110249.1 | - | 1 | 14 | |||
BC122529.1 | - | 1 | 267 | 40012183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
