siRNA Details
Gene Symbol: ERMARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278531.1 | NP_001265460.1 | 7 | 653 | ||
NM_001278532.1 | NP_001265461.1 | 6 | 484 | |||
NM_001278533.1 | NP_001265462.1 | 7 | 653 | |||
NM_018341.2 | NP_060811.1 | 7 | 653 | |||
XM_011535938.2 | XP_011534240.1 | 7 | 653 | |||
XM_011535939.2 | XP_011534241.1 | 7 | 1743 | |||
XM_011535940.2 | XP_011534242.1 | 7 | 1552 | |||
XM_017011027.1 | XP_016866516.1 | 7 | 1493 | |||
XM_017011028.1 | XP_016866517.1 | 7 | 818 | |||
XM_017011029.1 | XP_016866518.1 | 7 | 578 | |||
XM_017011030.1 | XP_016866519.1 | 6 | 402 | |||
XM_017011031.1 | XP_016866520.1 | 6 | 467 | |||
XM_017011032.1 | XP_016866521.1 | 7 | 1738 | |||
XM_017011033.1 | XP_016866522.1 | 7 | 1547 | |||
GenBank mRNA | AK001369.1 | - | 6 | 514 | ||
AK002014.1 | - | 7 | 650 | |||
AK294090.1 | - | 6 | 481 | |||
BC044812.1 | - | 7 | 637 | |||
CA396814.1 | - | 5 | 333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
