siRNA Details
Gene Symbol: ERMARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278531.1 | NP_001265460.1 | 15 | 1520 | ||
NM_001278532.1 | NP_001265461.1 | 14 | 1351 | |||
NM_001278533.1 | NP_001265462.1 | 15 | 1520 | |||
NM_018341.2 | NP_060811.1 | 15 | 1520 | |||
XM_011535938.2 | XP_011534240.1 | 14 | 1490 | |||
XM_011535939.2 | XP_011534241.1 | 15 | 2610 | |||
XM_011535940.2 | XP_011534242.1 | 15 | 2419 | |||
XM_017011027.1 | XP_016866516.1 | 15 | 2360 | |||
XM_017011028.1 | XP_016866517.1 | 15 | 1685 | |||
XM_017011029.1 | XP_016866518.1 | 15 | 1445 | |||
XM_017011030.1 | XP_016866519.1 | 14 | 1269 | |||
XM_017011031.1 | XP_016866520.1 | 14 | 1334 | |||
XM_017011032.1 | XP_016866521.1 | 15 | 2605 | |||
XM_017011033.1 | XP_016866522.1 | 15 | 2414 | |||
GenBank mRNA | AK001369.1 | - | 14 | 1381 | ||
AK002014.1 | - | 15 | 1517 | |||
AK021520.1 | - | 5 | 1584 | |||
AK294090.1 | - | 14 | 1348 | |||
BC044812.1 | - | 15 | 1504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
