siRNA Details
Gene Symbol: ZSWIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020928.1 | NP_065979.1 | 6 | 1612 | ||
XM_017009677.1 | XP_016865166.1 | 4 | 563 | |||
GenBank mRNA | AB046797.1 | - | 2 | 198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
