siRNA Details
Gene Symbol: UNC13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080421.2 | NP_001073890.2 | 20 | 2368 | ||
XM_011527810.2 | XP_011526112.1 | 19 | 2483 | |||
XM_011527811.2 | XP_011526113.1 | 20 | 2513 | |||
XM_017026502.1 | XP_016881991.1 | 20 | 2513 | |||
GenBank mRNA | AB028955.2 | - | 20 | 2366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
