siRNA Details
Gene Symbol: PXDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012293.2 | NP_036425.1 | 3 | 393 | ||
XM_005264707.3 | XP_005264764.1 | 3 | 381 | |||
XM_011510396.1 | XP_011508698.1 | 3 | 150 | |||
XM_011510397.2 | XP_011508699.1 | 3 | 310 | |||
GenBank mRNA | AF200348.1 | - | 3 | 352 | ||
BC030834.1 | - | 4 | 508 | |||
BC098579.1 | - | 3 | 288 | |||
BG706125.1 | - | 4 | 513 | |||
D86983.1 | - | 3 | 352 | |||
EF090903.1 | - | 3 | 352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
