siRNA Details
Gene Symbol: H19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002196.2 | 5 | 1758 | |||
NR_131223.1 | 5 | 1752 | ||||
NR_131224.1 | 5 | 649 | ||||
GenBank mRNA | AK092486.1 | - | 6 | 1209 | ||
AK123560.1 | - | 5 | 484 | |||
BC013067.1 | - | 5 | 600 | |||
BC023213.1 | - | 5 | 1070 | |||
BC098439.1 | - | 5 | 1118 | |||
BC106078.1 | - | 2 | 142 | |||
BC110657.1 | - | 5 | 1357 | |||
CN424819.1 | - | 5 | 653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
