siRNA Details
Gene Symbol: H19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002196.2 | 5 | 2005 | |||
NR_131223.1 | 5 | 1999 | ||||
NR_131224.1 | 5 | 896 | ||||
GenBank mRNA | AK092486.1 | - | 6 | 1456 | ||
AK123560.1 | - | 5 | 731 | |||
AK311497.1 | - | 2 | 488 | |||
BC013067.1 | - | 5 | 847 | |||
BC023213.1 | - | 5 | 1317 | |||
BC098439.1 | - | 5 | 1365 | |||
BC106078.1 | - | 2 | 389 | |||
BC110657.1 | - | 5 | 1604 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
