siRNA Details
Gene Symbol: H19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002196.2 | 3, 4 | 1609 | |||
NR_131223.1 | 3, 4 | 1609 | ||||
NR_131224.1 | 3, 4 | 500 | ||||
GenBank mRNA | AK092486.1 | - | 4, 5 | 1060 | ||
AK123560.1 | - | 3, 4 | 335 | |||
BC013067.1 | - | 3, 4 | 451 | |||
BC023213.1 | - | 3, 4 | 921 | |||
BC098439.1 | - | 3, 4 | 969 | |||
BC110657.1 | - | 3, 4 | 1208 | |||
BE730693.1 | - | 3, 4 | 487 | |||
CN424819.1 | - | 3, 4 | 504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
