siRNA Details
Gene Symbol: RIMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB002338.1 | - | 20 | 4239 | ||
AB045726.1 | - | 20 | 4482 | |||
BC151853.1 | - | 20 | 4263 | 8860502 | ||
BC152435.1 | - | 20 | 4239 | 8862556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
