siRNA Details
Gene Symbol: ZC3H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015168.1 | NP_055983.1 | 15 | 4998 | ||
XM_017026530.1 | XP_016882019.1 | 14 | 6436 | |||
XM_017026531.1 | XP_016882020.1 | 13 | 4779 | |||
GenBank mRNA | AB028987.2 | - | 15 | 4998 | ||
AK074282.1 | - | 1 | 912 | |||
AK125279.1 | - | 7 | 2810 | |||
AL050155.1 | - | 1 | 1986 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
