siRNA Details
Gene Symbol: C16orf95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195124.1 | NP_001182053.1 | 7 | 934 | ||
NM_001195125.1 | NP_001182054.1 | 5 | 750 | |||
NM_001256917.1 | NP_001243846.1 | 5 | 754 | |||
GenBank mRNA | AK026130.1 | - | 5 | 746 | ||
AL136762.2 | - | 1 | 635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
